Learn More
TNFRSF11B Monoclonal Antibody (5A11), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515726
329.34 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
MA5-15726 targets TNFRSF11B in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15726 immunogen is purified recombinant fragment of human TNFRSF11B expressed in E. Coli. MA5-15726 detects TNFRSF11B which has a predicted molecular weight of approximately 60kDa.
The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
Especificaciones
TNFRSF11B | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
O00300 | |
TNFRSF11B | |
Purified recombinant fragment of human TNFRSF11B expressed in E. Coli. | |
RUO | |
4982 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
5A11 | |
Unconjugated | |
TNFRSF11B | |
LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b; tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin) | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.