Learn More
TR2/HVEM Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ TR2/HVEM Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-710-PABX
Descripción
TNF receptor like-2 (TR2) (also known as herpesvirus entry mediator (HVEM)) is expressed in multiple human tissues and cell lines and shows a constitutive and relatively high expression in peripheral T and B lymphocytes, and monocytes. It is a member of TNF receptor family. TR2/HVEM is a receptor for herpes virus. It also acts as a receptor for LIGHT (also known as HVEM ligand), a member of TNF family. TR2 protein inhibits a mixed lymphocyte reaction-mediated proliferation suggesting that the receptor and/or its ligand play a role in T cell stimulation.Host Species: Mouse
Clone: 122
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human TR2 protein
Molecular Weight: 32kDa
Positive Control: Tonsil
Cellular Localization: Secreted protein
Recommended for:
- Immunofluorescence
- Flow Cytometry
- Function Blocking
Note:
USA: RUO; Int'l: RUO
Especificaciones
TR2/HVEM Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Infectious Disease | |
Human | |
IgG1 |
Flow Cytometry, Immunofluorescence | |
122 | |
Unconjugated | |
Recombinant human TR2 protein | |
RUO | |
Primary | |
Concentrated |