missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Mouse anti-Human, Clone: 70525, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
Marca: R&D Systems MAB626-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Immunohistochemistry, Neutralization.Especificaciones
TRANCE/TNFSF11/RANK L | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
8600 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Immunohistochemistry, Neutralization | |
70525 | |
Immunohistochemistry 8-25 ug/mL, Neutralization 0.8-2.5 ug/mL | |
O14788 | |
TNFSF11 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TRANCE/TNFSF11/RANK L Gly136-Asp317 (Ala194Gly) Accession # O14788 | |
25 μg | |
Primary | |
Detects human TRANCE/TNFSF11/RANK L in direct ELISAs. No cross-reactivity with recombinant mouse TRANCE is observed. | |
Human | |
Purified |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido