Learn More
Novus Biologicals™ Transferrin Native Protein
Highly purified. Generating reliable and reproducible results.
Marca: Novus Biologicals™ NBP1-96856
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Native Transferrin. Source: Serum The Transferrin Native Protein has been validated for the following applications: Western Blot, Dot Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.Especificaciones
Q06AH7 | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
Transferrin Protein | |
Human |
Dot Blot, Immunocytochemistry/Immunofluorescence | |
7018 | |
4°C in the dark. | |
Multi-step |