Learn More
Novus Biologicals™ Transferrin Native Protein
Highly purified. Generating reliable and reproducible results.
Marca:  Novus Biologicals™ NBP1-96856
Código de producto. 18002217
  • 296.00€ / 1 miligramo
Explore más ofertas especiales
Descripción
Descripción
Native Transferrin. Source: Serum The Transferrin Native Protein has been validated for the following applications: Western Blot, Dot Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.
Especificaciones
Especificaciones
Q06AH7
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Transferrin Protein
Human
Dot Blot, Immunocytochemistry/Immunofluorescence
7018
4°C in the dark.
Multi-step