missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trastuzumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Marca: Invitrogen MA542305
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Trastuzumab with the brand name Herceptin, is an FDA-approved drug for HER2-targeted therapy for breast cancer. It is a recombinant humanized monoclonal antibody (IgG1 kappa). Trastuzumab binds to HER2 receptor present on the surface of HER2-positive tumour cells and suppresses the cells growth and proliferation.Especificaciones
Trastuzumab | |
Recombinant Monoclonal | |
Unconjugated | |
Herceptin; Metastatic lymph node gene 19 | |
Human ERBB2 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
2.6 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 |