missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin alpha-1B Monoclonal Antibody (OTI3G3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA526265
372.01 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
The microtubules are intracellular dynamic polymers made up of evolutionarily conserved polymorphic alpha/beta-tubulin heterodimers and a large number of microtubule-associated proteins (MAPs). Microtubules have their intrinsic polarity; highly dynamic plus ends and less dynamic minus ends. Microtubules are required for vital processes in eukaryotic cells including mitosis, meiosis, maintenance of cell shape and intracellular transport. Microtubules are also necessary for movement of cells by means of flagella and cilia. In mammalian tissue culture cells microtubules have their minus ends anchored in microtubule organizing centers (MTOCs).The GTP (guanosintriphosphate) molecule is an essential for tubulin heterodimer to associate with other heterodimers to form microtubule. In vivo, microtubule dynamics vary considerably. Microtubule polymerization is reversible and a populations of microtubules in cells are on their minus ends either growing or shortening - this phenomenon is called dynamic instability of microtubules. The alpha-tubulin (relative molecular weight around 50 kDa) is globular protein that exists in cells as part of soluble alpha/beta-tubulin dimer or it is polymerized into microtubules. In different species it is coded by multiple tubulin genes that form tubulin classes (in human 6 genes).
Especificaciones
Tubulin alpha-1B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P68363 | |
TUBA1B | |
Full length human recombinant protein of TUBA1B produced in HEK293T cell | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3G3 | |
Unconjugated | |
TUBA1B | |
alpha tubulin; alpha-tubulin 2; Alpha-tubulin isotype M-alpha-2; alpha-tubulin ubiquitous; Detyrosinated tubulin alpha-1B chain; K-ALPHA-1; RGD1565476; Tuba1b; Tuba2; tubulin alpha 1b; tubulin alpha-1B chain; Tubulin alpha-2 chain; tubulin alpha-ubiquitous chain; Tubulin K-alpha-1; tubulin, alpha 1B; tubulin, alpha 2; tubulin, alpha, ubiquitous | |
Mouse | |
Affinity Chromatography | |
RUO | |
10376 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Tubulin alpha-1B Monoclonal Antibody (OTI3G3), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido