missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tunicamycin, Tocris Bioscience™
Click to view available options
Cantidad:
10 mg
Descripción
Tunicamycin is an antibiotic; inhibits GlcNAc phosphotransferase (GPT). Blocks the formation of N-glycosidic linkages by inhibiting the first step in glycoprotein synthesis. Activity induces ER stress and causes G1 arrest; can be used to induce autophagy. Tunicamycin contains four main components as follows: Homolog A, n=8, C37H60N4O16, molecular weight = 816.90 Homolog B, n=9, C38H62N4O16, molecular weight = 830.93 Homolog C, n=10, C39H64N4O16, molecular weight = 844.95 Homolog D, n=11, C40H66N4O16, molecular weight = 858.99 The composition of this product will vary from batch to batch and can be found on the relevant certificate of analysis.
Especificaciones
Nombre del producto químico o material | Tunicamycin |
Tipo | Tunicamycin |
CAS | 11089-65-9 |
Fórmula molecular | C39H64N4O16 (tunicamycin C, n=10) |
Sinónimo | Tunicamycin from Streptomyces sp. |
Formula Weight (peso de la fórmula) | 844.95 |
Porcentaje de pureza | >98% |
Cantidad | 10 mg |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'