missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550813
Detalles adicionales : Peso : 0.01000kg
Descripción
The antibody detects endogenous levels of total NME8 / TXNDC3 protein.
This gene encodes a protein with an N-terminal thioredoxin domain and three C-terminal nucleoside diphosphate kinase (NDK) domains, but the NDK domains are thought to be catalytically inactive. The sea urchin ortholog of this gene encodes a component of sperm outer dynein arms, and the protein is implicated in ciliary function. Mutations in this gene are implicated in primary ciliary dyskinesia type 6.Especificaciones
TXNDC3 | |
Polyclonal | |
Unconjugated | |
NME8 | |
1700056P15Rik; CILD6; epididymis secretory protein Li 99; HEL-S-99; NM23-H8; NME/NM23 family member 8; Nme8; spermatid-specific thioredoxin-2; spermatid-specific thioredoxin-2 protein; sperm-specific thioredoxin 2; SPTRX2; sptrx-2; thioredoxin domain containing 3 (spermatozoa); thioredoxin domain-containing protein 3; Txndc3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51314 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N427 | |
NME8 | |
Fusion protein corresponding to residues near the C terminal of human NME/NM23 family member 8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |