missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYRO3 Mouse anti-Human, Clone: OTI5B7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF500415
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, and cytoskeleton reorganization.Especificaciones
TYRO3 | |
Monoclonal | |
Unconjugated | |
TYRO3 | |
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif | |
Mouse | |
Affinity Chromatography | |
RUO | |
7301 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
OTI5B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q06418 | |
TYRO3 | |
Full length human recombit protein of human TYRO3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |