Learn More
DuPont™ Sterile Tyvek™ IsoClean Overboots
Use overboots in cleanroom class ISO 5 and higher environments to maintain quality. Sterile Tyvek™ IsoClean Overboots have a reinforced seam and repel water-based liquids and aerosols. Clean laundered and gamma-sterilised.
236.98€
Especificaciones
Material | Tyvek |
---|---|
Color | White |
Desechables | Single-use |
Tipo de suela | Slip-resistant |
Tipo de costura | Reinforced seam protection |
Código de producto | Marca | Precio | Cantidad y disponibilidad | ||||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Precio | Cantidad y disponibilidad | ||||||
15483123
|
DuPont™
D14246800 |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
|||||||
15493123
|
DuPont™
D14246825 |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
|||||||
Descripción
Tyvek™ IsoClean- Sterile overboot in white
- Fixation ties
- Tunnelled elastics at ankles and gripper slip-retardant full sole
- Composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Offers an excellent barrier against fine particles and fibers (down to micron in size) and is ultra-low-linting
- Silicon non-added
- Garments and accessories made of Tyvek™ IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments
Disposable Overboots
- This accessory is not CE certified as chemical protective clothing
- Clean laundered and gamma-sterilised
- Suitable for use in cleanroom class ISO 5 and higher
- Sterility assurance level (SAL) of 10-6
- Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
- Full traceability on all sterilized apparel with certificates of sterility available
Especificaciones
Tyvek | |
Single-use | |
Reinforced seam protection |
White | |
Slip-resistant | |
100 |