Learn More
UBE1L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530675
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: IMR32. Predicted reactivity: Mouse (89%), Rat (90%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
UBA6 (UBE1L2, MOP-4) is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins. UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility. UBA6 is highly expressed in human monocytes.Especificaciones
UBE1L2 | |
Polyclonal | |
Unconjugated | |
UBA6 | |
4930542H01; 5730469D23Rik; AU021846; AW124799; E1-L2; Monocyte protein 4; MOP4; MOP-4; RGD1308324; UBA6; UBA6, ubiquitin-activating enzyme E1; Ube1l2; ubiquitin like modifier activating enzyme 6; ubiquitin-activating enzyme 6; ubiquitin-activating enzyme E1-like 2; ubiquitin-activating enzyme E1-like protein 2; ubiquitin-like modifier activating enzyme 6; ubiquitin-like modifier-activating enzyme 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55236 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
A0AVT1 | |
UBA6 | |
Recombinant fragment corresponding to a region within amino acids 147 and 368 of Human FLJ10808 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |