missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567631
333.35 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
Predicted to react with Mouse samples.
Receptor for netrin required for axon guidance. Mediates axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. Mediates apoptosis by activating DAPK1. In the absence of NTN1, activates DAPK1 by reducing its autoinhibitory phosphorylation at Ser-308 thereby increasing its catalytic activity.
Especificaciones
UNC5B | |
Polyclonal | |
Unconjugated | |
UNC5B | |
6330415E02Rik; A630020F16; C. elegans; D10Bwg0792e; Netrin receptor UNC5B; p53RDL1; p53-regulated receptor for death and life protein 1; Protein unc-5 homolog 2; protein unc-5 homolog B; transmembrane receptor Unc5H2; unc5 homolog 2; unc-5 homolog 2; unc-5 homolog B; unc-5 homolog B (C. elegans); unc-5 netrin receptor B; Unc5b; UNC5H2; UNQ1883/PRO4326 | |
Rabbit | |
affinity chromatography | |
RUO | |
107449, 219699 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IZJ1, Q8K1S3 | |
UNC5B | |
Synthesized peptide derived from human UNC5B. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
UNC5B Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido