Learn More
Deltalab™ Recipientes universales de poliestireno
Polystyrene containers with screw caps are commonly used within the lab for various purposes, primarily for the storage and transportation of liquid samples, reagents and solutions. The screw cap provides a secure seal to prevent leakage or contamination during handling or storage.
Marca: Deltalab™ 309222.O
Detalles adicionales : Peso : 0.01000kg
Descripción
- EtO sterilised
- Individually packaged
- Lightweight and durable
- Resistant to breakage
- Suitable for a wide range of applications
- Transparent to allow for easy visual inspection of contents
Especificaciones
Recipiente universal | |
No | |
Estéril | |
30 mL | |
Polypropylene | |
Blanco | |
Sample collection, storage and transportation | |
No label | |
1 in. |
Yes | |
Poliestireno | |
1 oz. | |
White | |
Tapón de rosca | |
Universal polystyrene container | |
3 in. | |
400/paq. | |
25 mm |