Learn More
Lovibond™ ValidCheck Chlorine
Creating a Primary Chlorine Standard has never been easier! This self-contained ValidCheck Chlorine kit will allow for preparation of 100 mL of 1.5 mg/L NIST traceable primary chlorine standard without need for complicated dilutions or the additional use of pipettes or glassware.
Marca:  Lovibond™ 48105510
Código de producto. 17884892
  • 32.00€ / cada uno
Fecha estimada de envío: 24-01-2025
para ver el stock
Explore más ofertas especiales
Descripción
  • Ready-to-use solutions
  • Exactly adjusted concentrations
  • Adapted to special applications
  • Multistandards with top-up solution
Especificaciones
Especificaciones
1.5 mg/L Chlorine Standard