missing translation for 'onlineSavingsMsg'
Learn More

VAP-1 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

214.00€ - 1796.00€

Especificaciones

Antígeno VAP-1
Concentración 0.1-1.0 mg/mL
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17855113
Ver documentos
Invitrogen
500-P326-50UG
50μg
214.00€
50 microgramos
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17815323
Ver documentos
Invitrogen
500-P326-100UG
100 μg
269.00€
100 microgramos
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
17895653
Ver documentos
Invitrogen
500-P326-1MG
1 mg
1796.00€
1 miligramo
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

AA Seq. of rec. protein: GRGGDGGEPS QLPHCPSVSP SAQPWTHPGQ SQLFADLSRE ELTAVMRFLT QRLGPGLVDA AQARPSDNCV FSVELQLPPK AAALAHLDRG SPPPAREALA IVFFGRQPQP NVSELVVGPL PHPSYMRDVT VERHGGPLPY HRRPVLFQEY LDIDQMIFNR ELPQASGLLH HCCFYKHRGR NLVTMTTAPR GLQSGDRATW FGLYYNISGA GFFLHHVGLE LLVNHKALDP ARWTIQKVFY QGRYYDSLAQ LEAQFEAGLV NVVLIPDNGT GGSWSLKSPV PPGPAPPLQF YPQGPRFSVQ GSRVASSLWT FSFGLGAFSG PRIFDVRFQG ERLVYEISLQ EALAIYGGNS PAAMTTRYVD GGFGMGKYTT PLTRGVDCPY LATYVDWHFL LESQAPKTIR DAFCVFEQNQ GLPLRRHHSD LYSHYFGGLA ETVLVVRSMS TLLNYDYVWD TVFHPSGAIE IRFYATGYIS SAFLFGATGK YGNQVSEHTL GTVHTHSAHF KVDLDVAGLE NWVWAEDMVF VPMAVPWSPE HQLQRLQVTR KLLEMEEQAA FLVGSATPRY LYLASNHSNK WGHPRGYRIQ MLSFAGEPLP QNSSMARGFS WERYQLAVTQ RKEEEPSSSS VFNQNDPWAP TVDFSDFINN ETIAGKDLVA WVTAGFLHIP HAEDIPNTVT VGNGVGFFLR PYNFFDEDPS FYSADSIYFR GDQDAGACEV NPLACLPQAA ACAPDLPAFS HGGFSHN. Sandwich ELISA: To detect Hu VAP-1 by sandwich ELISA (using 100 uL/well), a conc. of 0.5-2.0 ug/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Rabbit Anti-Hu VAP-1 (500-P326BT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of rec. Hu VAP-1. WB: To detect Hu VAP-1 by WB analysis this antibody can be used at a conc. of 0.1-0.2 ug/mL. When used in conjunction with compatible secondary reagents, the detection limit for rec. Hu VAP-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
Especificaciones

Especificaciones

VAP-1
ELISA, Western Blot
Unconjugated
Rabbit
Human
Q16853
8639
E.coli-derived Recombinant Human VAP-1
Antigen affinity chromatography
AOC3
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97
AOC3
Primary
-20°C
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
VAP-1 Polyclonal Antibody, PeproTech®, Invitrogen™

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado