missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT2 Monoclonal Antibody (CL2921), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA531378
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: CGFIHEDELD EETGDITQNY INYGTTKSYG ATTQANGGWP SGWEKKEEFV QGEVQDSHSY KDRVDY
VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.Especificaciones
VGLUT2 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8BLE7, Q9JI12, Q9P2U8 | |
SLC17A6 | |
Recombinant protein corresponding to Human SLC17A6 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry | |
CL2921 | |
Unconjugated | |
SLC17A6 | |
2900073D12Rik; differentation-associated Na-dependent inorganic phosphate cotransporter; differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; Dnpi; SLC17A6; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGLUT2 | |
Mouse | |
Protein A | |
RUO | |
140919, 57084, 84487 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |