Learn More
Invitrogen™ H-2Kd/H-2Dd Monoclonal Antibody (34-1-2S)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA518008
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-18008 targets H-2KdDd in CT and FACS applications and shows reactivity with Mouse samples. The MA5-18008 immunogen is bDF splenocytes.
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Especificaciones
| H-2Kd/H-2Dd | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P01895, P01896, P01897, P01899, P01900, P01901, P03991, P04223, P14426, P14427, P14428 | |
| H2-D1, H2-K1 | |
| BDF splenocytes. | |
| 250 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Cytotoxicity Assay, Flow Cytometry | |
| 34-1-2S | |
| Unconjugated | |
| H2-D1 | |
| class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, D-B alpha chain; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2K1; H2-K1; H2KD; histocompatibility 2, D region locus 1; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-D-q alpha-chain; MHC H2-K transplantation antigen; Q5k protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 14964, 14972 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |