missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (VIM/1937R), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-08936PE
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifikationer
| Vimentin | |
| Monoclonal | |
| PE | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| Recombinant full-length human Vimentin protein (Uniprot: P08670) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| VIM/1937R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Bovine, Canine | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto