missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIPR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534301
Detalles adicionales : Peso : 0.01000kg
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey, Marmoset, Rabbit (94%) Hamster, Pig (81%).
The Vasoactive Intestinal Polypeptide Receptor 1 (VPAC1) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.Especificaciones
VIPR1 | |
Polyclonal | |
Unconjugated | |
VIPR1 | |
AV071699; HVR1; II; Pacap receptor; PACAP type II receptor; PACAP-R2; PACAP-R-2; pituitary adenylate cyclase activating polypeptide receptor, type II; pituitary adenylate cyclase-activating polypeptide type II receptor; Pvr2; RATVASREC; RDC1; type 1 vasoactive intestinal peptide receptor; V1RG; VAPC1; vasoactive intestinal peptide receptor 1; vasoactive intestinal polypeptide receptor 1; Vasopressive intestinal peptide receptor; VASREC; VIP and PACAP receptor 1; VIP receptor 1; VIP receptor subtype 1; VIP receptor, type I; VIP1 receptor; VIPR; Vipr1; VIP-R1; VIP-R-1; VIRG; VPAC1; VPAC1 receptor; VPAC1R; VPCAP1R | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P32241 | |
VIPR1 | |
Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human VIP Receptor 1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |