missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vismodegib, MedChemExpress
Description
Vismodegib (GDC-0449) is an orally active hedgehog pathway inhibitor with an IC50 of 3 nM. Vismodegib also inhibits P-gp, ABCG2 with IC50 values of 3.0 μM and 1.4 μM, respectively.
Specifications
Specifications
| Nombre del producto químico o material | Vismodegib |
| CAS | 879085-55-9 |
| Color | Off-White |
| Notas de grado de pureza | Research |
| Fórmula molecular | C19H14Cl2N2O3S |
| Cantidad | 50 mg |
| Sinónimo | GDC-0449 |
| Información de solubilidad | DMSO : 50 mg/mL (118.68 mM; Need ultrasonic) ∣Ethanol : 3.33 mg/mL (7.90 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(C1=CC=C(C=C1Cl)S(=O)(C)=O)NC2=CC=C(C(C3=NC=CC=C3)=C2)Cl |
| Peso molecular (g/mol) | 421.3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?