Learn More
Medicom WeePro Labo Sterile Integral Coverall with Boots, Type 5/6
Click to view available options
Tamaño:
Medium
Large
X-Large
2X-Large
Descripción
- New material MICROMIUM™, light and very low contaminant
- Cleanroom apparel compatible from ISO class 4
- Chemical splash suit, Category III Type 5/6
- Double packaging fully sealed
- Apparel sterilized by gamma irradiation
Especificaciones
Tipo de producto | Cleanroom Coveralls |
Tamaño | Medium |
Sexo | Unisex |
Material | Microporous Fabric |
Color | White |
Desechables | Single-use |
Estilo de cintura | Elastic (Stitched-in) |
Estilo de tobillo | Covered Elastic |
Estilo de manguito | Elastic |
Estilo de cuello | Hooded |