missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilm's Tumor 1 Rabbit, Clone: WT1/1434R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 7490-RBM4-P0
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Wilm's Tumor 1 | |
Nuclear and Cytoplasmic | |
WT1/1434R | |
Unconjugated | |
NP_001185480 | |
Rabbit | |
47-55kDa | |
Protein A/G Purified | |
RUO | |
7490 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
WT1 | |
Recombinant human full-length WT1 protein | |
K562 cells. Wilm s Tumor, mesothelioma or fetal kidney. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |