Learn More
Assistant™ Wooden Tongue Depressor
Descripción
- High-quality birch wood
- Not waxed
- Polished without rough edges
- Without splinters
- Odorless and tasteless
- Food-safe
Especificaciones
Especificaciones
| Material | Birch Wood |
| Certificaciones/conformidad | CE |
| Color | Natural |
| Dimensiones (Largo x Ancho x Alto) | 150 x 18 x 1.6 mm (±0.15 mm) |
| Esterilidad | No estéril |