Learn More
Circonio, solución estándar de AAS, Specpure™, Zr 1000 μg/ml, Thermo Scientific Chemicals
Código de producto. 11306389
Click to view available options
Cantidad:
100 mL
500 mL
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Número MDL MFCD00011303
Especificaciones
Nombre del producto químico o material Zirconium, AAS standard solution
Concentración ZrCl2O in 5% HCl
Nombre de nota Zr 1000μg/mL
Cantidad 500 mL
Número UN UN1789
Grado Specpure™
Forma física Líquido