missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF187 293T Cell Overexpression Lysate (Denatured), Abnova
Marca: Abnova H00007741-T01.100uL
Detalles adicionales : Peso : 0.00010kg
Descripción
Western Blotting
Especificaciones
Western Blot | |
ZNF187 293T Cell Transient Overexpression Lysate(Denatured) | |
37.5 | |
pCMV-ZNF187 full-length | |
100 μL | |
Human |
293T | |
BC013962.2 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-ZNF187 antibody by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |