missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZO-1 Monoclonal Antibody (ZO1-1A12), FITC
GREENER_CHOICE

Código de producto. 15722476
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
15722476 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15722476

Marca: Invitrogen™ 339111

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Species reactivity includes human (Caco-2 Cell line) and dog (MDCK cell line). Based on sequence homology, reactivity with other species is likely but has not been confirmed. This monoclonal antibody detects ZO-1alpha+ and ZO-1alpha- isoforms. The antibody is specific for ZO-1 proteins; cross-reactivity with the related ZO-2 protein has not been observed. For this FITC conjugate the excitation (EX)/emission (EM) are 494/520 (nm).

ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ZO-1
Aplicaciones ELISA, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon ZO1-1A12
Concentración 0.5 mg/mL
Conjugado FITC
Formulación PBS with 1% BSA, 50% glycerol and 0.1% sodium azide; pH 7.4
génica TJP1
N.º de referencia del gen O97758, Q07157
Alias de gen DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1
Símbolos de los genes TJP1
Especie del huésped Mouse
Inmunógeno Human recombinant ZO-1 fusion protein encompassing amino acids 334-634.
Método de purificación Affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 403752, 7082
Especies diana Human, Canine
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.