CAS RN 1008768-41-9
CAS RN 1008768-41-9
IUPAC Name:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ TM2 TEAD inhibitor
Potent and reversible inhibitor of TEAD mediated Hippo/Yap signalling