CAS RN 103-25-3
CAS RN 103-25-3
IUPAC Name:
methyl 3-phenylpropanoate
Sinónimos:
methyl phenylpropionate
Peso molecular (g/mol):
164.2
Fórmula molecular:
C10H12O2
InChi Key:
RPUSRLKKXPQSGP-UHFFFAOYSA-N
SMILES:
COC(=O)CCC1=CC=CC=C1
1
–
1
de
1
resultados
1
3-Fenilpropionato de metilo, 98 %, Thermo Scientific Chemicals
CAS: 103-25-3 Fórmula molecular: C10H12O2 Peso molecular (g/mol): 164.204 Número MDL: MFCD00017209 Clave InChI: RPUSRLKKXPQSGP-UHFFFAOYSA-N Sinónimo: methyl 3-phenylpropionate,methyl hydrocinnamate,benzenepropanoic acid, methyl ester,3-phenylpropionic acid methyl ester,methyl benzenepropanoate,hydrocinnamic acid, methyl ester,methyl dihydrocinnamate,methyl beta-phenylpropionate,methyl b-phenylpropionate,unii-111lc1gi10 PubChem CID: 7643 Nombre IUPAC: 3-fenilpropanoato de metilo SMILES: COC(=O)CCC1=CC=CC=C1