missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dichloro-2-phenylcyclopropane, 97%, Thermo Scientific™

Product Code. 11445003
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11445003

Brand: Thermo Scientific Alfa Aesar A12279.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2415-80-7
Molecular Formula C9H8Cl2
Molecular Weight (g/mol) 187.063
MDL Number MFCD00001272
InChI Key WLWFQGXZIDYWQF-UHFFFAOYSA-N
Synonym 2,2-dichlorocyclopropyl benzene, 1,1-dichloro-2-phenylcyclopropane, 2,2-dichlorocyclopropyl-benzene, benzene, 2,2-dichlorocyclopropyl, cyclopropane, 1,1-dichloro, 2-phenyl, pubchem18679, acmc-1clka, ksc491m2n, wlwfqgxzidywqf-uhfffaoysa, 2,2-dichloro-cyclopropyl-benzene
PubChem CID 95349
IUPAC Name (2,2-dichlorocyclopropyl)benzene
SMILES C1C(C1(Cl)Cl)C2=CC=CC=C2

Specifications

Density 1.208
Boiling Point 105°C to 106°C (12 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.551
Quantity 25 g
Beilstein 1860351
Formula Weight 187.07
Percent Purity 97%
Chemical Name or Material 1,1-Dichloro-2-phenylcyclopropane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.