missing translation for 'onlineSavingsMsg'
Learn More
1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Thermo Scientific™ Product Code.: 10470944

1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Thermo Scientific™

Product Code. 10470944
25 g
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 10470944

Brand: Thermo Scientific Maybridge SB01820EB

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Chemical Identifiers

CAS 37989-92-7
Molecular Formula C15H13FO
Molecular Weight (g/mol) 228.266
MDL Number MFCD00236612
InChI Key ZXIQZWXVSBJPTC-UHFFFAOYSA-N
Synonym 1-2'-fluoro-1,1'-biphenyl-4-yl propan-1-one, 1-2'-fluoro 1,1'-biphenyl-4-yl propan-1-one, 1-4-2-fluorophenyl phenyl propan-1-one, 4'-2-fluorophenyl propiophenone, 4-2-fluorophenyl propiophenone, 1-propanone,1-2'-fluoro 1,1'-biphenyl-4-yl, maybridge3_006207, 2-fluoro-4'-propanoylbiphenylShow MoreShow Less
PubChem CID 2737490
IUPAC Name 1-[4-(2-fluorophenyl)phenyl]propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula FC6H4C6H4C(O)CH2CH3
Quantity 25 g
Formula Weight 228.26
Percent Purity 97%
Chemical Name or Material 1-(2'-Fluoro[1, 1'-biphenyl]-4-yl)propan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.