Learn More
Hamilton™ 100 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfills the general liquid handling requirements of manufacturing and research laboratories.
Brand: Hamilton™ CAL80630
Additional Details : Peso : 0.02300kg
Description
- The 700 series is the original Hamilton syringe.
- This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
- The 700 series remains the industry standard.
- The plungers and barrels are not interchangeable or replaceable.
- Dispenses volumes up to 100 μL.
Specifications
Not Autoclavable | |
22s G | |
0.0059 in. | |
0.0283 in. | |
1.49 mm | |
0.72 mm | |
122°F | |
50°F | |
10°C | |
100 μL | |
Sharp, Bevel, Curved, Non-coring |
Borosilicate Glass | |
0.061 in. | |
0.15 mm | |
0.305 in. | |
7.75 mm | |
2 in | |
50°C | |
710 N | |
1 | |
51 mm | |
Cemented |