missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiene, 90%, Tech., mixture of isomers

Product Code. 10295371 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10295371

Brand: Thermo Scientific Acros 346670050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-46-1 | C6H10 | 82.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-46-1
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00009297,MFCD00009297
InChI Key APPOKADJQUIAHP-CIIODKQPSA-N
Synonym unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
PubChem CID 643786
IUPAC Name (2Z,4E)-hexa-2,4-diene
SMILES C\C=C\C=C/C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow
Density 0.7200g/mL
Boiling Point 82°C
Flash Point -8°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCH=CHCH3
Refractive Index 1.45
Quantity 5 g
Beilstein 01, 254
Specific Gravity 0.72
Solubility Information Solubility in water: insoluble
Formula Weight 82.14
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2, 4-Hexadiene, tech. 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Hexadiene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.