missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitro-N,N-di-n-propylaniline, 97%, Thermo Scientific™

Product Code. 15460889
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15460889 5 g 5g
15470889 250 mg 250mg
15450889 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 15460889

Brand: Thermo Scientific Alfa Aesar H63921.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6216-91-7
Molecular Formula C12H17ClN2O2
Molecular Weight (g/mol) 256.73
InChI Key OYJJPFCGGNTTTO-UHFFFAOYSA-N
Synonym 2-chloro-4-nitro-nn-dipropylaniline, 4-nitro-2-chloro-n,n-di-n-propyl aniline, benzenamine,2-chloro-4-nitro-n,n-dipropyl, 2-chloro-4-nitrophenyl dipropylamine, 2-chloro-4-nitro-phenyl-dipropyl-amine
PubChem CID 138688
IUPAC Name 2-chloro-4-nitro-N,N-dipropylaniline
SMILES CCCN(CCC)C1=C(C=C(C=C1)[N+](=O)[O-])Cl

Specifications

Quantity 5 g
UN Number UN2811
Formula Weight 256.73
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-4-nitro-N,N-di-n-propylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.