missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodo-4-nitrotoluene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.
Solubility
Soluble in toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.
Solubility
Soluble in toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.

Specifications
Specifications
Quantity | 100 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in toluene. |
Formula Weight | 263.03 |
Percent Purity | 97% |
Chemical Name or Material | 2-Iodo-4-nitrotoluene |
Product Suggestions
Los clientes que vieron este artículo también vieron
Viewing 1-5 of 10
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Iodo-4-nitrotoluene, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction