missing translation for 'onlineSavingsMsg'
Learn More

2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one, 98%, Thermo Scientific™

Product Code. 11348185
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11348185 1 g 1g
11358185 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11348185

Brand: Thermo Scientific Alfa Aesar L08473.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20348-09-8
Molecular Formula C7H6N2O2
Molecular Weight (g/mol) 150.137
MDL Number MFCD00006697
InChI Key ANHQLUBMNSSPBV-UHFFFAOYSA-N
Synonym 2h-pyrido 3,2-b 1,4 oxazin-3 4h-one, 2h-pyrido 3,2-b-1,4-oxazin-3 4h-one, 4h-pyrido 3,2-b 1,4 oxazin-3-one, 2h,3h,4h-pyrido 3,2-b 1,4 oxazin-3-one, 2h,4h-pyrido 3,2-b 1,4 oxazin-3-one, 2h,4h-pyridino 2,3-e 1,4-oxazin-3-one, pyridoboxazinone, acmc-20ahw7, 2h-pyrido 3,2-b 1,4 oxazin-3-one
PubChem CID 88499
IUPAC Name 4H-pyrido[3,2-b][1,4]oxazin-3-one
SMILES C1C(=O)NC2=C(O1)C=CC=N2

Specifications

Melting Point 204°C to 208°C
Quantity 1 g
Beilstein 1074010
Formula Weight 150.14
Percent Purity 98%
Chemical Name or Material 2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.