missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Benzothiazolyl)aniline, 96%

Product Code. 15415897
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15415897 250 mg 250mg
15405897 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15415897

Brand: Thermo Scientific Alfa Aesar H33103.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41230-21-1 | C13H10N2S | 226.297 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41230-21-1
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.297
MDL Number MFCD00579111
InChI Key MKMRZZJJDIBAJF-UHFFFAOYSA-N
Synonym 3-benzothiazol-2-yl-phenylamine, 3-1,3-benzothiazol-2-yl aniline, 3-benzothiazol-2-ylphenylamine, benzenamine,3-2-benzothiazolyl, cambridge id 5250055, 3-2-benzothiazolyl aniline, 3-benzothiazol-2-yl aniline, 3-benzo d thiazol-2-yl aniline, 3-benzothiazol-2-yl benzenamine, 3-1,3-benzothiazol-2-yl phenyl amine
PubChem CID 720999
IUPAC Name 3-(1,3-benzothiazol-2-yl)aniline
SMILES C1=CC=C2C(=C1)N=C(S2)C3=CC(=CC=C3)N

Specifications

Melting Point 135°C to 137°C
Quantity 250 mg
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 226.3
Percent Purity 96%
Chemical Name or Material 3-(2-Benzothiazolyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.