missing translation for 'onlineSavingsMsg'
Learn More

3-[2-Fluoro-5-(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Product Code. 15467985
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15467985 250 mg 250mg
15457985 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15467985

Brand: Thermo Scientific Alfa Aesar H32071.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-39-8
Molecular Formula C10H8F4O2
Molecular Weight (g/mol) 236.166
MDL Number MFCD09025356
InChI Key AGDPEKNRXANVOW-UHFFFAOYSA-N
Synonym 3-2-fluoro-5-trifluoromethyl phenyl propionic acid, 3-2-fluoro-5-trifluoromethyl phenyl propanoic acid
PubChem CID 46737486
IUPAC Name 3-[2-fluoro-5-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)CCC(=O)O)F

Specifications

Melting Point 61°C to 64°C
Quantity 250 mg
Formula Weight 236.16
Percent Purity 97%
Chemical Name or Material 3-[2-Fluoro-5-(trifluoromethyl)phenyl]propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.