Learn More
3-Methoxy-5-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
Product Code. 15445457
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15445457 1 g 1g
15455457 5 g 5g
2 options
This item is not returnable. View return policy
Product Code. 15445457
Brand: Thermo Scientific Alfa Aesar H32703.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 916420-92-3
Molecular Formula C9H6ClF3O2
Molecular Weight (g/mol) 238.59
MDL Number MFCD09025410
InChI Key IHMFMHOCMZSKNI-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl benzoyl chloride, 5-methoxy-3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-methoxy-5-trifluoromethyl
PubChem CID 46737576
IUPAC Name 3-methoxy-5-(trifluoromethyl)benzoyl chloride
SMILES COC1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl
Specifications
Refractive Index 1.4915
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 238.59
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzoyl chloride