missing translation for 'onlineSavingsMsg'
Learn More

Ácido 4-acetilbenzoico, +98 %, Thermo Scientific Chemicals

40.40€ - 392.96€

Chemical Identifiers

CAS 586-89-0
Fórmula molecular C9H8O3
Peso molecular (g/mol) 164.16
Número MDL MFCD00002561
Clave InChI QBHDSQZASIBAAI-UHFFFAOYSA-N
Sinónimo benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
PubChem CID 11470
Nombre IUPAC ácido 4-acetilbenzoico
SMILES CC(=O)C1=CC=C(C=C1)C(=O)O
View More Specs

Products 3
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
11482278
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A15239.03
1 g
40.40€
1 g
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
11492278
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A15239.06
5 g
117.59€
5 g
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
11402288
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A15239.14
25 g
392.96€
25 g
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El ácido 4-acetilbenzoico se empleaba en la preparación de un derivado de éster de paclitaxel.

Solubilidad
Soluble en agua.

Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes.
Specifications

Chemical Identifiers

586-89-0
164.16
QBHDSQZASIBAAI-UHFFFAOYSA-N
11470
CC(=O)C1=CC=C(C=C1)C(=O)O
C9H8O3
MFCD00002561
benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
ácido 4-acetilbenzoico

Specifications

4-Acetylbenzoic acid
208°C to 212°C
C9H8O3
2207355
Soluble in water.
CC(=O)C1=CC=C(C=C1)C(=O)O
164.16
164.16
586-89-0
Odorless
MFCD00002561
benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
QBHDSQZASIBAAI-UHFFFAOYSA-N
ácido 4-acetilbenzoico
11470
≥98%
Videos
Safety and Handling

Safety and Handling

Declaración de GHS H
H315-H319-H335
Provoca irritación cutánea.
Provoca irritación ocular grave.
Puede irritar las vías respiratorias.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335.

missing translation for 'einecsNumber' : 209-588-5

missing translation for 'tsca' : No

Recommended Storage : Temperaturas ambientes

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.