missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-5-fluorobenzoic acid, 98%, Thermo Scientific™

Product Code. 15410969
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15410969 25 g 25g
15490959 1 g 1g
15400969 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15410969

Brand: Thermo Scientific Alfa Aesar H63965.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177480-81-8
Molecular Formula C7H3BrClFO2
Molecular Weight (g/mol) 253.45
MDL Number MFCD00236589
InChI Key TZFKKLGCVKORLS-UHFFFAOYSA-N
Synonym 4-bromo-2-chloro-5-fluoro-benzoic acid, benzoic acid, 4-bromo-2-chloro-5-fluoro, pubchem1361, acmc-1c1y0, 4-bromo-2-chloro-5-fluorobenzoicacid, 2-chloro-4-bromo-5-fluorobenzoic acid
PubChem CID 2736225
IUPAC Name 4-bromo-2-chloro-5-fluorobenzoic acid
SMILES OC(=O)C1=CC(F)=C(Br)C=C1Cl

Specifications

Color White
Quantity 25 g
Formula Weight 253.45
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-chloro-5-fluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.