Learn More
4-Bromo-3,3,4,4-tetrafluoro-1-buteno, 98 %, Thermo Scientific™
37.10€ - 121.00€
Identificadores de productos químicos
CAS | 18599-22-9 |
---|---|
Fórmula molecular | C4H3BrF4 |
Peso molecular (g/mol) | 206.97 |
Número MDL | MFCD00042189 |
Clave InChI | GVCWGFZDSIWLMO-UHFFFAOYSA-N |
Sinónimo | 4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene |
PubChem CID | 87721 |
Nombre IUPAC | 4-bromo-3,3,4,4-tetrafluorobut-1-eno |
SMILES | FC(F)(Br)C(F)(F)C=C |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
18599-22-9 | |
206.97 | |
GVCWGFZDSIWLMO-UHFFFAOYSA-N | |
87721 | |
FC(F)(Br)C(F)(F)C=C |
C4H3BrF4 | |
MFCD00042189 | |
4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene | |
4-bromo-3,3,4,4-tetrafluorobut-1-eno |
Especificaciones
4-Bromo-3,3,4,4-tetrafluoro-1-butene | |
1.627 | |
None | |
BrCF2CF2CH=CH2 | |
MFCD00042189 | |
4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene | |
FC(F)(Br)C(F)(F)C=C | |
206.97 | |
206.97 |
18599-22-9 | |
55°C to 57°C | |
C4H3BrF4 | |
1.354 | |
1856181 | |
GVCWGFZDSIWLMO-UHFFFAOYSA-N | |
4-bromo-3,3,4,4-tetrafluorobut-1-eno | |
87721 | |
98% |
Seguridad y manipulación
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
missing translation for 'einecsNumber' : 242-440-8
missing translation for 'tsca' : Yes
Recommended Storage : Temperaturas ambientes
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.