missing translation for 'onlineSavingsMsg'
Learn More

4-cis-Fluoro-L-prolinamide hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11362239
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 426844-23-7
Molecular Formula C5H10ClFN2O
Molecular Weight (g/mol) 168.596
MDL Number MFCD07368567
InChI Key YIEACUBSPQGCLW-MMALYQPHSA-N
Synonym 2s,4s-4-fluoropyrrolidine-2-carboxamide hydrochloride, 4-cis-fluoro-l-prolinamide hydrochloride, 2s,4s-4-fluoropyrrolidine-2-carboxamide hcl, cis-4-fluoro-l-prolinamide hydrochloride, 2s,4s-4-fluoro-2-pyrrolidinecarboxamide hydrochloride, 4s-4-fluoro-l-prolinamide-hydrogen chloride 1/1, 2s,4s-2-carbamoyl-4-fluoropyrrolidine hydrochloride salt, 2s,4s-4-fluoropyrrolidine-2-carboxamide monohydrochloride, 2s,4s-2-aminocarbonyl-4-fluoropyrrolidine hydrochloride
PubChem CID 45158917
IUPAC Name (2S,4S)-4-fluoropyrrolidine-2-carboxamide;hydrochloride
SMILES C1C(CNC1C(=O)N)F.Cl

Specifications

Melting Point 231°C to 236°C (decomposition)
Quantity 1 g
Optical Rotation −45 (c=1 in Water)
Formula Weight 168.59
Percent Purity 98%
Chemical Name or Material 4-cis-Fluoro-L-prolinamide hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-cis-Fluoro-L-prolinamide hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.