missing translation for 'onlineSavingsMsg'
Learn More

5-[4-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole, 95%, Thermo Scientific™

Product Code. 10626874
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10626874 1 g Amber Glass Bottle 1g
11320687 250 mg Amber Glass Bottle 250mg
2 options
This item is not returnable. View return policy

Product Code. 10626874

Brand: Thermo Scientific Maybridge CC50908DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 362529-03-1
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
MDL Number MFCD09025866
InChI Key RWTXHMRWDVQEOV-UHFFFAOYSA-N
Synonym 5-4-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-4-bromomethyl phenyl-3-methyl, 1,2,4-oxadiazole,5-4-bromomethyl phenyl-3-methyl
PubChem CID 18374480
IUPAC Name 5-[4-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
SMILES CC1=NOC(=N1)C2=CC=C(C=C2)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.1
Percent Purity 95%
Chemical Name or Material 5-[4-(Bromomethyl)phenyl]-3-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.