missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoylthiophene-2-boronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15489745
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15489745 1 g 1g
15499745 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15489745

Brand: Thermo Scientific Alfa Aesar H52353.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Carbamoylthiophene-2-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957344-92-2
Molecular Formula C11H16BNO3S
Molecular Weight (g/mol) 253.12
MDL Number MFCD17215214
InChI Key SQHNPVGOAXWLAM-UHFFFAOYSA-N
Synonym 5-carbamoylthiophene-2-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene-2-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophene-2-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxa-borolan-2-yl-thiophene-2-carboxylic acid amide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophene-2-carboxylic acid amide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl thiophene-2-carboxylic acid amide
PubChem CID 57589940
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxamide
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(S1)C(N)=O

Specifications

Quantity 1 g
Formula Weight 253.13
Percent Purity 96%
Chemical Name or Material 5-Carbamoylthiophene-2-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.