Learn More
Hamilton™ 50 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfi the general liquid handling requirements of manufacturing and research laboratories.
Marca:  Hamilton™ CAL80530
 Ver más versiones de este producto
Código de producto. 17824342
  • 349.00€ / cada uno
Explore más ofertas especiales
Descripción
Descripción
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 50 μL.
Especificaciones
Especificaciones
Not Autoclavable
22s G
0,0059 in
0,0283 in
1.03 mm
0.72 mm
122 °F
50 °F
10 °C
50 μl
Afilada, biselada, curva, sin perforación
Stainless Steel
0,041 in
0.15 mm
0,305 in
7.75 mm
2 in
50 °C
705 N
1
51 mm
Cementada