Learn More
Hamilton™ 50 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfi the general liquid handling requirements of manufacturing and research laboratories.
Marca: Hamilton™ CAL80901
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The 700 series is the original Hamilton syringe.
- This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
- The 700 series remains the industry standard.
- The plungers and barrels are not interchangeable or replaceable.
- Dispenses volumes up to 50 μL.
Especificaciones
Not Autoclavable | |
22s G | |
0,0059 in | |
0,0283 in | |
1.03 mm | |
0.72 mm | |
Jeringa Microliter | |
50 °C | |
705 N | |
1 | |
51 mm | |
Cementada |
Stainless Steel | |
0,041 in | |
0.15 mm | |
0,305 in | |
7.75 mm | |
2 in | |
122 °F | |
50 °F | |
10 °C | |
50 μl | |
Romo |