Learn More
Hamilton™ 50 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfi the general liquid handling requirements of manufacturing and research laboratories.
Marca:  Hamilton™ CAL7655-01
 Ver más versiones de este producto
Código de producto. 17864212
  • 287.00€ / cada uno
Explore más ofertas especiales
Descripción
Descripción
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 50 μL.
Especificaciones
Especificaciones
Autoclavable
No Needle Included
0,305 in
7.75 mm
Jeringa Microliter
115°C
705 RN
1
No Needle Included
Extraíble
Stainless Steel
0,041 in
1.03 mm
No Needle Included
239 °F
50 °F
10 °C
50 μl
No Needle Included