Learn More
Merck Amicon™ Stirred Cell Seal Kits
Use with solutions up to 10% macrosolute concentration. Amicon™ Stirred Cell Seal Kit includes 3 silicone gaskets and O-rings. The Amicon stirred cell model provides high flow rates and is capable of rapid concentration or salt removal / concentration in the same unit.
Marca: Merck UFSC200SL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Amicon™ Stirred Cell Seal Kit (3 each o-ring and gasket)Especificaciones
Autoclavable | |
28.7 cm2 | |
Amicon™ Stirred Cell 200 mL Seal Kit (3 Ea. of o-ring and gasket) | |
Ultrafiltration Equipment | |
15.5 cm | |
63.5 mm (filter) |
Silicone | |
Protein desalting, purification and concentration | |
3 Ea. gaskets and O-rings | |
200 mL | |
63.5 mm filter |