Learn More
AMP, solución tampón de 0,2 M, pH 9,5, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar J63988.AK
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
124-68-5 | |
Solución de tampón de 0,2 m | |
Líquido | |
C4H11NO | |
MFCD00008051 | |
CBTVGIZVANVGBH-UHFFFAOYSA-N | |
2-amino-2-metilpropan-1-ol |
Incolora | |
9.5 | |
250 mL | |
Odorless | |
2-Amino-2-methyl-1-propanol | |
CC(C)(N)CO | |
89.14 |