missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGR2 Monoclonal Antibody (10E2), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA516244
340.01 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).
Especificaciones
AGR2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% sodium azide | |
O88312, O95994 | |
Agr2 | |
A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody. | |
100 μg | |
Primary | |
Bovine, Canine, Equine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
10E2 | |
Unconjugated | |
Agr2 | |
AG2; AG-2; AGR2; agr2 protein; Agr2h; AII; AII type II arginase; anterior gradient 2; anterior gradient 2 homolog; anterior gradient 2 homologue; anterior gradient 2, protein disulphide isomerase family member; anterior gradient homolog 2; anterior gradient protein 2 homolog; ARG2; arginase 2; arginase II; arginase type II; arginase, type II; arginase-2, mitochondrial; AU022422; epididymis secretory protein Li 116; Gob4; Gob-4; HAG-2; HEL-S-116; HPC8; kidney arginase; kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; mAG-2; nonhepatic arginase; Non-hepatic arginase; PDIA17; protein disulfide isomerase family A, member 17; protein Gob-4; secreted cement gland homolog; secreted cement gland protein XAG-2 homolog; Type II arginase; UNQ515/PRO1030; XAG-2 | |
Mouse | |
Protein G | |
RUO | |
100053075, 10551, 23795, 298961, 415112, 482333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
AGR2 Monoclonal Antibody (10E2), Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido